missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ QSY™ 7 C5-Maleimide

Catalog No. Q10257
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q10257 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q10257 Supplier Molecular Probes™ Supplier No. Q10257

QSY™ 7 C5-maleimide

QSY™ 7 C5-maleimide is a nonfluorescent acceptor dye (absorption maximum ∼560 nm) for preparation of FRET probes via thiol modification.

Specifications

Quantity 5 mg
Product Type Maleimide
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye QSY™ 7
Label Type Quencher Dyes
Product Line QSY

For Research Use Only. Not for use in diagnostic procedures.