Learn More
Qorpak™ Unlined Polypropylene Caps
Polypropylene caps are known for good impact strength, durability, cost-effectiveness, thermal resistance, and pliability.
Description
- Autoclavable
- Well-suited for many dry products
- Add a PTFE disc liner to the unlined polypropylene cap for better leak resistance when autoclaving
Chemical Identifiers
| White | |
| Non-sterile | |
| Wide Mouth Bottle |
| Polypropylene | |
| Reusable |
Specifications
| White | |
| 1300 Caps/Cs. | |
| Non-sterile | |
| Reusable | |
| Wide Mouth Bottle |
| Polypropylene | |
| Bulk | |
| 53-400 | |
| Ribbed Cap | |
| Not Autoclavable |