Learn More
Qorpak™ ValuLine F217 and PTFE Lined Polypropylene Caps
Help to keep bottle or jar contents free of odor and taste interference
Supplier: Qorpak™ 239089
Description
- Made of white polypropylene with PTFE facing
- F217 liner backing consisting of low density foam core tightly sandwiched between two layers of LDPE
- Resilient with strong resistance to chemicals and moisture
Chemical Identifiers
| White | |
| Bulk | |
| F217, PTFE | |
| Lined Cap (Ribbed) | |
| Not Autoclavable |
| Polypropylene | |
| Non-sterile | |
| Reusable | |
| Wide mouth bottle |
Specifications
| White | |
| 760 Cs. | |
| Non-sterile | |
| 70-400 | |
| Lined Cap (Ribbed) | |
| Not Autoclavable |
| Polypropylene | |
| Bulk | |
| F217, PTFE | |
| Reusable | |
| Wide mouth bottle |