missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ ValuLine F217 and PTFE Lined Polypropylene Caps

Catalog No. p-4603621
Change view
Click to view available options
Neck Finish:
28-400
48-400
70-400
89-400
No. per Case:
1700 Cs.
456 Cs.
5000 Cs.
760 Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
239080 28-400 5000 Cs.
239085 48-400 1700 Cs.
239089 70-400 760 Cs.
239090 89-400 456 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 239080 Supplier Qorpak™ Supplier No. 239080
Only null left

Help to keep bottle or jar contents free of odor and taste interference

  • Made of white polypropylene with PTFE facing
  • F217 liner backing consisting of low density foam core tightly sandwiched between two layers of LDPE
  • Resilient with strong resistance to chemicals and moisture

Specifications

Color White
Material Polypropylene
No. per Case 5000 Cs.
Packaging Bulk
Sterility Non-sterile
Liner Material F217, PTFE
Neck Finish 28-400
Disposable Reusable
Product Type Lined Cap (Ribbed)
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Show More Show Less