missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Clear Straight Sided Round Bottles with Green Thermoset F217 and PTFE Caps

Catalog No. 02992436
Change view
Click to view available options
Capacity (Metric):
60 mL
120 mL
480 mL
960 mL
Quantity:
12/Cs.
24/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
02992436 120 mL 24/Cs.
02992428 60 mL 24/Cs.
02992415 480 mL 12/Cs.
02992406 960 mL 12/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02992436 Supplier Qorpak™ Supplier No. GLC01634

Designed without shoulders for maximum storage capacity

  • Provides content visibility
  • Closure (Green thermoset (phenolic)) provides the chemical compatibility and most temperature tolerant
  • Foam backed PTFE liners provides resilience for tight seal with low moisture transmission rate

Analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.

Specifications

Capacity 4 oz., 120 mL
Capacity (Metric) 120 mL
Color Clear
Diameter (English) 2.36 in.
Material Glass
Quantity 24/Cs.
Sterility Non-sterile
Closure Color Green
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Height (Metric) 68 mm
Height (English) 2.68 in.
Closure Liner F217, PTFE
Closure Material Thermoset
Closure Size 58-400 mm
Diameter (Metric) 60 mm
Closure Type Screw Cap
Autoclavable Not Autoclavable
Capacity (English) 4 oz.
Shape Straight-sided
Product Type Wide Mouth Bottle
Show More Show Less