missing translation for 'onlineSavingsMsg'
Learn More

QKI Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501557053
Encompass
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501557053 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501557053 Supplier Bethyl Laboratories Inc Supplier No. A300183AT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to

QKI belongs to a family of RNA-binding proteins that have an HNRNPK (MIM 600712) homology (KH) domain embedded in a 200-amino acid region called the GSG domain. Other members of this family include SAM68 (KHDRBS1; MIM 602489) and SF1 (MIM 601516) (Chen and Richard, 1998 [PubMed 9671495]). QKI proteins regulate RNA splicing, export of target RNAs from the nucleus, translation of proteins, and RNA stability (Lauriat et al., 2008 [PubMed 17918747]).

Specifications

Antigen QKI
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene QKI
Gene Accession No. Q96PU8, Q9QYS9
Gene Alias Protein quaking; HqkI; Hqk; QKI; HKQ
Gene Symbols Qk, QKI
Host Species Rabbit
Immunogen C-terminus of isoform 5
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19317, 9444
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less