missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Qdot™ 585 ITK™ Streptavidin Conjugate Kit

Catalog No. Q10011MP
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q10011MP 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q10011MP Supplier Molecular Probes™ Supplier No. Q10011MP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Qdot™ 585 ITK™ Streptavidin Conjugate Kit

The Qdot™ 585 ITK™ Streptavidin Conjugate Kit provides the Qdot™ streptavidin conjugate as 250μL of a 2μM solution. Unlike the Qdot™ streptavidin conjugates, the Qdot™ ITK™ streptavidin conjugates have the streptavidin covalently attached to the inner amphiphilic coating without a PEG linker. These Qdo™ conjugates can be used for binding biotinylated probes. The kit also includes 30 mL of Qdot™ incubation buffer.

Specifications

Conjugate Qdot 585
Quantity 1 Kit
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Room Temperature
Product Line ITK, Qdot
Product Type Conjugate Kit

For Research Use Only. Not for use in diagnostic procedures.