missing translation for 'onlineSavingsMsg'
Learn More

QC Standard 7 For Environmental Analysis, SPEX CertiPrep™

Catalog No. p-219569
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QC7 125 mL
QC7500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. QC7 Supplier SPEX CertiPrep Group LLC Supplier No. QC7
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
K1000μg/mL
Ag100μg/mL
Al100μg/mL
B100μg/mL
Ba100μg/mL
Na100μg/mL
Si50μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material ICP Quality Control Standard 7
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 5% HNO3/tr F
Quantity 125 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Components in 5% HNO3/tr. HF:
Al    •100μg/mL
Ba    •100μg/mL
B    •100μg/mL
K    •1000μg/mL
Si    •50μg/mL
Ag    •100μg/mL
Na    •100μg/mL

Product intended for laboratory use only