missing translation for 'onlineSavingsMsg'
Learn More

QC Standard 21 For Environmental Analysis, SPEX CertiPrep™

Catalog No. QC21
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QC21 125 mL
QC21500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. QC21 Supplier SPEX CertiPrep Group LLC Supplier No. QC21

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
As100μg/mL
Be100μg/mL
Ca100μg/mL
Cd100μg/mL
Co100μg/mL
Cr100μg/mL
Cu100μg/mL
Fe100μg/mL
Li100μg/mL
Mg100μg/mL
Mn100μg/mL
Mo100μg/mL
Ni100μg/mL
Pb100μg/mL
Sb100μg/mL
Se100μg/mL
Sr100μg/mL
Ti100μg/mL
Tl100μg/mL
V100μg/mL
Zn100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material ICP Quality Control Standard 21
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride
Quantity 125 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Each component at 100μg/mL in 5% HNO3/tr. Tartaric Acid/tr. HF:, Antimony, Arsenic, Beryllium, Cadmium, Calcium, Chromium, Cobalt, Copper, Iron, Lead, Lithium, Magnesium, Manganese, Molybdenum, Nickel, Selenium, Strontium, Thallium, Titanium, Vanadium, Zinc

Product intended for laboratory use only