missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ QC Standard 21, Ricca Chemical

Catalog No. RV010724100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010724100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010724100 Supplier Ricca Chemical Company Supplier No. RV010724100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

100 ppm: As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn /nManufac

Specifications

Chemical Name or Material Verispec™ QC Standard 21, 100 ppm: As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb,
Name Note Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Packaging Natural LDPE Bottle
CAS 7697-37-2, 7778-39-4, 7697-37-2
CAS Min % 2, 0.02
CAS Max % 2, 0.02
Density 1.09
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Calibration Standard
Quantity 100 mL
Grade High Purity
Show More Show Less