missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QA-1b Monoclonal Antibody (6A8.6F10.1A6)
GREENER_CHOICE

Catalog No. PIMA524801
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524801 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524801 Supplier Invitrogen™ Supplier No. MA524801
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

QA-1b Monoclonal Antibody for Western Blot, Flow, IP

The Qa-1b gene is the mouse homolog of the human HLA-E, a non-classical major histocompatibility complex class Ib protein encoded by the T23 gene within the T region of the H-2 complex. Qa-1b plays a crucial role in immune regulation by serving as a ligand for murine CD94/NKG2A natural killer (NK) cell inhibitory receptors. It primarily binds to a nonameric peptide known as the Qa-1 determinant modifier (Qdm), derived from the leader sequence of other MHC class Ia molecules. In mouse melanoma cells, particularly the B16-F10 line, Qa-1b is overexpressed, and its knockdown has been observed to inhibit cell proliferation and migration, while also promoting cell cycle arrest at the G0/G1 phase and facilitating apoptosis via the Ras-Raf-MAPK signaling pathway. As such, Qa-1b serves as an oncogenic factor in melanoma models, offering potential therapeutic pathways for cancer treatment. Its non-polymorphic nature contributes to its conserved function across various mouse strains, with only two dominant allelic variants, Qa-1a and Qa-1b. Despite sharing a 73% homology with human HLA-E, differences in peptide repertoires and binding affinities highlight its unique role in murine immune responses.

Specifications

Antigen QA-1b
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6A8.6F10.1A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene H2-T23
Gene Accession No. P06339
Gene Alias 37b; 37c; H-2 class I histocompatibility antigen, D-37 alpha chain; H2-Q1; H2-Qa1; H-2T23; H2-T23; histocompatibility 2, T region locus 23; MHC class I antigen Qa1; MHC classIb Qa1; Qa1; Qa-1; Qa-1(b); Qed-1; T18c; T18c(37); T23b; T23d
Gene Symbols H2-T23
Host Species Mouse
Immunogen Synthetic peptide from the alpha-2 domain of Qa-1b.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15040
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less