missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Q Sepharose™ High Performance Chromatography Media

Catalog No. 45002945
Change view
Click to view available options
Quantity:
75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002945 75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002945 Supplier Cytiva Supplier No. 17101401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for high intermediate and polishing steps in downstream processing

  • Composed of crosslinked agarose beads with mean diameter of 34μm, modified with quaternary (Q) strong anion exchange groups
  • Hydrophilic nature of base matrix ensures low levels of non-specific binding
  • High resolution, high capacity separation with high recovery
  • Chemical stability for effective CIP/sanitization
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

Specifications

Quantity 75 mL
For Use With (Application) For Intermediate PurifiCatIon and Polishing AppliCatIons
Stability Stable in commonly used aqueous Buffers: 8 M Urea, 6 M Guanidine HCl, 70% Ethanol, 1 M NaOH, 1 M Acetic Acid, 30% Isopropanol, 30% Acetonitrile, 2% SDS
Storage Requirements 4°C to 30°C
Product Type Q Sepharose High Performance Media
Flow Rate 30 to 150 cm/Hoμr
Particle Size 24 to 44 μm
Height (English) Bed 3.93 in.
Height (Metric) Bed 10 cm
Ionic Capacity 0.14 to 0.20 mM Cl-/mL
Ion Exchange Type Strong AnIon Exchanger
Max. Pressure <3 bar
Matrix 6% Cross-Linked Agarose
pH Range 1 to 14
Storage Buffer 20% Ethanol
Ligand Type Quaternary Amine
Show More Show Less