missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvic Aldehyde Dimethyl Acetal 97.0+%, TCI America™
Supplier: TCI America P1264100ML
Specifications
| Pyruvic Aldehyde Dimethyl Acetal | |
| 141°C | |
| MFCD00008758 | |
| 1993 | |
| ULVSHNOGEVXRDR-UHFFFAOYSA-N | |
| 1,1-dimethoxypropan-2-one | |
| 80650 | |
| ≥97.0% (GC) |
| 6342-56-9 | |
| C5H10O3 | |
| 100 mL | |
| 1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal | |
| CC(=O)C(OC)OC | |
| 118.132 | |
| 118.13 | |
| Liquid |
Chemical Identifiers
| 6342-56-9 | |
| 118.132 | |
| ULVSHNOGEVXRDR-UHFFFAOYSA-N | |
| 80650 | |
| CC(=O)C(OC)OC |
| C5H10O3 | |
| MFCD00008758 | |
| 1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal | |
| 1,1-dimethoxypropan-2-one |
Safety and Handling
TSCA : Yes