missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvic Aldehyde Dimethyl Acetal 97.0+%, TCI America™
$148.25 - $733.49
Chemical Identifiers
| CAS | 6342-56-9 |
|---|---|
| Molecular Formula | C5H10O3 |
| Molecular Weight (g/mol) | 118.132 |
| MDL Number | MFCD00008758 |
| InChI Key | ULVSHNOGEVXRDR-UHFFFAOYSA-N |
| Synonym | 1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal |
| PubChem CID | 80650 |
| IUPAC Name | 1,1-dimethoxypropan-2-one |
| SMILES | CC(=O)C(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P126425ML
|
TCI America
P126425ML |
25 mL |
Each for $148.25
|
|
|||||
|
P1264100ML
|
TCI America
P1264100ML |
100 mL |
Each for $304.18
|
|
|||||
|
P1264500ML
|
TCI America
P1264500ML |
500 mL |
Each for $733.49
|
|
|||||
Chemical Identifiers
| 6342-56-9 | |
| 118.132 | |
| ULVSHNOGEVXRDR-UHFFFAOYSA-N | |
| 80650 | |
| CC(=O)C(OC)OC |
| C5H10O3 | |
| MFCD00008758 | |
| 1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal | |
| 1,1-dimethoxypropan-2-one |
Specifications
| 6342-56-9 | |
| C5H10O3 | |
| 25 mL | |
| 1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal | |
| CC(=O)C(OC)OC | |
| 118.132 | |
| 118.13 | |
| Liquid |
| 141°C | |
| MFCD00008758 | |
| 1993 | |
| ULVSHNOGEVXRDR-UHFFFAOYSA-N | |
| 1,1-dimethoxypropan-2-one | |
| 80650 | |
| ≥97.0% (GC) | |
| Pyruvic Aldehyde Dimethyl Acetal |
Safety and Handling
TSCA : Yes