missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrrolidine Hydriodide 98.0+%, TCI America™
Supplier: TCI America P24865G
Specifications
| Pyrrolidine Hydriodide | |
| C4H10IN | |
| Pyrrolidinium Iodide | |
| C1CCNC1.I | |
| 199.035 | |
| 199.04 | |
| Crystalline Powder |
| 45361-12-4 | |
| 5 g | |
| DMFMZFFIQRMJQZ-UHFFFAOYSA-N | |
| pyrrolidine;hydroiodide | |
| 12196047 | |
| ≥98.0% (N,T) |
Chemical Identifiers
| 45361-12-4 | |
| 199.035 | |
| Pyrrolidinium Iodide | |
| pyrrolidine;hydroiodide |
| C4H10IN | |
| DMFMZFFIQRMJQZ-UHFFFAOYSA-N | |
| 12196047 | |
| C1CCNC1.I |
Safety and Handling
TSCA : No