missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine Hydriodide 98.0+%, TCI America™

Catalog No. p-7135582
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24861G 1 g
P24865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24861G Supplier TCI America Supplier No. P24861G
Only null left

Chemical Identifiers

CAS 45361-12-4
Molecular Formula C4H10IN
Molecular Weight (g/mol) 199.035
InChI Key DMFMZFFIQRMJQZ-UHFFFAOYSA-N
Synonym Pyrrolidinium Iodide
PubChem CID 12196047
IUPAC Name pyrrolidine;hydroiodide
SMILES C1CCNC1.I

Specifications

Quantity 1 g
Formula Weight 199.04
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrrolidine Hydriodide