missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine-3-carboxylic acid, 98%

Catalog No. AC459110010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459110010 1 g
AC459110050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459110010 Supplier Thermo Scientific Chemicals Supplier No. 459110010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59378-87-9 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59378-87-9
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
InChI Key JAEIBKXSIXOLOL-UHFFFAOYSA-N
Synonym 3-pyrrolidinecarboxylic acid, 3-pyrrolidine carboxylic acid, beta-proline, 3-pyrrolidinecarboxylicacid, pyrrolidine-3-carboxylicacid, dl-pyrrolidine-3-carboxylic acid, beta-dl-proline, pubchem10439, acmc-209mcv, acmc-20a9vh
PubChem CID 3034645
IUPAC Name pyrrolidine-3-carboxylic acid
SMILES C1CNCC1C(=O)O

Specifications

Melting Point 182.0°C to 184.0°C
Elemental Analysis 50.86 to max. 53.47% min. Carbon 11.86 to max. 12.47% min. Nitrogen
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 115.13
Percent Purity 98%
Chemical Name or Material Pyrrolidine-3-carboxylic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes serious eye damage.
Precautionary Statement GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only