CAS RN 59378-87-9
IUPAC Name:
pyrrolidine-3-carboxylic acid
Synonyms:
pyrrolidine-3-carboxylic acid
Molecular Weight (g/mol):
115.13
Molecular Formula:
C5H9NO2
InChi Key:
JAEIBKXSIXOLOL-UHFFFAOYNA-N
SMILES:
OC(=O)C1CCNC1
1
–
1
of
1
results
1
Promotions Available
Pyrrolidine-3-carboxylic acid, 98%
Pyrrolidine-3-carboxylic acid, 98%
CAS: 59378-87-9 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.13 InChI Key: JAEIBKXSIXOLOL-UHFFFAOYSA-N Synonym: 3-pyrrolidinecarboxylic acid,3-pyrrolidine carboxylic acid,beta-proline,3-pyrrolidinecarboxylicacid,pyrrolidine-3-carboxylicacid,dl-pyrrolidine-3-carboxylic acid,beta-dl-proline,pubchem10439,acmc-209mcv,acmc-20a9vh PubChem CID: 3034645 IUPAC Name: pyrrolidine-3-carboxylic acid SMILES: C1CNCC1C(=O)O