missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine-1-carboximidamide hydroiodide, 97%, Thermo Scientific™

Catalog No. p-2705786
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
KM00683EB 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM00683EB Supplier Thermo Scientific Chemicals Supplier No. KM00683EB
Only null left

Chemical Identifiers

CAS 102392-83-6
Molecular Formula C5H12IN3
Molecular Weight (g/mol) 241.076
MDL Number MFCD00114285
InChI Key XPASGQSDLPDWNV-UHFFFAOYSA-N
Synonym pyrrolidine-1-carboximidamide hydroiodide, pyrrolidinecarboxamidine, iodide, c5h11n3.hi
PubChem CID 2794596
IUPAC Name pyrrolidine-1-carboximidamide;hydroiodide
SMILES C1CCN(C1)C(=N)N.I

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 241.07
Percent Purity 97%
Chemical Name or Material Pyrrolidine-1-carboximidamide hydroiodide
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.