missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carbonitrile, 99%

Catalog No. AAL1533003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1533003 1 g
AAL1533006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1533003 Supplier Thermo Scientific Chemicals Supplier No. L1533003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4513-94-4 | C5H4N2 | 92.10 g/mol

Pyrrole-2-carbonitrile is useful in addition reactions and in heterocycle formation. It acts as a heterocyclic building block in synthetic chemistry. Further, it is used in the preparation of 4-iodopyrrole-2-carbonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-2-carbonitrile is useful in addition reactions and in heterocycle formation. It acts as a heterocyclic building block in synthetic chemistry. Further, it is used in the preparation of 4-iodopyrrole-2-carbonitrile.

Solubility
Miscible with benzene, dichloromethane, tertrahydrofuran, dimethylformamide and acetonitrile.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 4513-94-4
Molecular Formula C5H4N2
Molecular Weight (g/mol) 92.10
MDL Number MFCD00234061
InChI Key BQMPGKPTOHKYHS-UHFFFAOYSA-N
Synonym pyrrole-2-carbonitrile, 2-cyanopyrrole, unii-6d25bmc1pp, 2-carbonitrilepyrrole, 6d25bmc1pp, cyanopyrrole, 2-cyano-pyrrol, pyrrole carbonitrile, 1h-pyrrolecarbonitrile, 1h-pyrrol-2-kohlenitrile
PubChem CID 138277
IUPAC Name 1H-pyrrole-2-carbonitrile
SMILES N#CC1=CC=CN1

Specifications

Density 1.086
Boiling Point 92°C to 94°C (2 mmHg)
Refractive Index 1.55
Quantity 1 g
UN Number UN3276
Beilstein 107047
Sensitivity Air and light sensitive
Solubility Information Miscible with benzene,dichloromethane,tertrahydrofuran,dimethylformamide and acetonitrile.
Formula Weight 92.1
Percent Purity 99%
Chemical Name or Material Pyrrole-2-carbonitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only