accessibility menu, dialog, popup

UserName

CAS RN 4513-94-4

IUPAC Name: 1H-pyrrole-2-carbonitrile
Synonyms: 2-cyanopyrrole
Molecular Weight (g/mol): 92.1
Molecular Formula: C5H4N2
InChi Key: BQMPGKPTOHKYHS-UHFFFAOYSA-N
SMILES: N#CC1=CC=CN1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)