missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carbonitrile, 97%

Catalog No. p-2692122
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC368350010 1 g Glass bottle
AC368350050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC368350010 Supplier Thermo Scientific Chemicals Supplier No. 368350010
Only null left

CAS: 4513-94-4 | C5H4N2 | 92.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4513-94-4
Molecular Formula C5H4N2
Molecular Weight (g/mol) 92.10
MDL Number MFCD00234061
InChI Key BQMPGKPTOHKYHS-UHFFFAOYSA-N
Synonym pyrrole-2-carbonitrile, 2-cyanopyrrole, unii-6d25bmc1pp, 2-carbonitrilepyrrole, 6d25bmc1pp, cyanopyrrole, 2-cyano-pyrrol, pyrrole carbonitrile, 1h-pyrrolecarbonitrile, 1h-pyrrol-2-kohlenitrile
PubChem CID 138277
IUPAC Name 1H-pyrrole-2-carbonitrile
SMILES N#CC1=CC=CN1

Specifications

Color Yellow to Orange
Boiling Point 115°C to 120°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5475 to 1.5505
Quantity 1 g
Formula Weight 92.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Pyrrole-2-carbonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only