missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUJIFILM Wako PYROSTAR™ ES-F 200 Test Kit
Endotoxin-specific and specially formulated to be unreactive to (1,3)- ß-D-glucan, Multi-test configuration designed for use with water bath, heat block or Toxinometer™
$1,158.25
Specifications
| Form | Lyophilized |
|---|---|
| Fill Volume | 5.2 mL |
| Includes | 4 x 5.2 mL vials of lysate and 1 vial of CSE (500 ng/vial) |
| Quantity | 200 Tests |
| Catalog Number | Mfr. No. | Sensitivity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sensitivity | Price | Quantity & Availability | |||||
|
12333005
|
FUJIFILM Wako
WPEK450015 |
0.015 EU/mL |
Each for $1,158.25
|
|
|||||
|
12333006
|
FUJIFILM Wako
WPEK450003 |
0.03 EU/mL |
Each for $1,158.25
|
|
|||||
|
12333007
|
FUJIFILM Wako
WPEK450006 |
0.06 EU/mL |
Each for $1,158.25
|
|
|||||
|
12333008
|
FUJIFILM Wako
WPEK450125 |
0.125 EU/mL |
Each for $1,158.25
|
|
|||||
|
12333009
|
FUJIFILM Wako
WPEK450025 |
0.25 EU/mL |
Each for $1,158.25
|
|
|||||
Description
- Endotoxin-specific reagent avoids false positive results from glucans
- Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
- Gel clot sensitivities range from 0.015 to 0.25 EU/mL
- Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
- Chances of interference are reduced due to high sensitivity
- Facilitates easier transition from gel clot to KTA
Specifications
| Lyophilized | |
| 5.2 mL | |
| 4 x 5.2 mL vials of lysate and 1 vial of CSE (500 ng/vial) | |
| 200 Tests |