Learn More
FUJIFILM Wako PYROSTAR™ ES-F 200 Test Kit
Endotoxin-specific and specially formulated to be unreactive to (1,3)- ß-D-glucan, Multi-test configuration designed for use with water bath, heat block or Toxinometer™
Supplier: FUJIFILM Wako WPEK450003
Description
- Endotoxin-specific reagent avoids false positive results from glucans
- Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
- Gel clot sensitivities range from 0.015 to 0.25 EU/mL
- Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
- Chances of interference are reduced due to high sensitivity
- Facilitates easier transition from gel clot to KTA
Chemical Identifiers
| Lyophilized | |
| 5.2 mL | |
| 4 x 5.2 mL vials of lysate and 1 vial of CSE (500 ng/vial) | |
| 200 Tests |
Specifications
| Lyophilized | |
| 4 x 5.2 mL vials of lysate and 1 vial of CSE (500 ng/vial) | |
| 0.03 EU/mL |
| 5.2 mL | |
| 200 Tests |