missing translation for 'onlineSavingsMsg'
Learn More

Pyromellitic Diimide 97.0+%, TCI America™

Catalog No. p-7134978
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11535G 5 g
P115325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11535G Supplier TCI America Supplier No. P11535G
Only null left

Chemical Identifiers

CAS 2550-73-4
Molecular Formula C10H4N2O4
Molecular Weight (g/mol) 216.152
MDL Number MFCD00005004
InChI Key UGQZLDXDWSPAOM-UHFFFAOYSA-N
Synonym pyromellitic diimide, pyromellitimide, pyromellitic acid diimide, benzo 1,2-c:4,5-c' dipyrrole-1,3,5,7 2h,6h-tetrone, unii-32xa0w314g, 1,2,4,5-benzenetetracarboxylic acid diimide, 1,2,4,5-benzenetetracarboxylic 1,2:4,5-diimide, benzene-1,2,4,5-tetracarboxylic diimide, pyrrolo 3,4-f isoindole-1,3,5,7-tetrone, pyrrolo 3,4-f isoindole-1,3,5,7 2h,6h-tetrone
PubChem CID 75696
IUPAC Name pyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
SMILES C1=C2C(=CC3=C1C(=O)NC3=O)C(=O)NC2=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 216.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyromellitic Diimide