accessibility menu, dialog, popup

UserName

OHNOONHO

CAS RN 2550-73-4

IUPAC Name: 1H,2H,3H,5H,6H,7H-pyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
Synonyms: 2H,6H-pyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
Molecular Weight (g/mol): 216.15
Molecular Formula: C10H4N2O4
InChi Key: UGQZLDXDWSPAOM-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2=CC3=C(C=C12)C(=O)NC3=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)