missing translation for 'onlineSavingsMsg'
Learn More

Pyromellitic diimide, 97%

CAS: 2550-73-4 | C10H4N2O4 | 216.152 g/mol

$83.68 - $273.62

Chemical Identifiers

CAS 2550-73-4
Molecular Formula C10H4N2O4
Molecular Weight (g/mol) 216.152
MDL Number MFCD00005004
InChI Key UGQZLDXDWSPAOM-UHFFFAOYSA-N
Synonym pyromellitic diimide, pyromellitimide, pyromellitic acid diimide, benzo 1,2-c:4,5-c' dipyrrole-1,3,5,7 2h,6h-tetrone, unii-32xa0w314g, 1,2,4,5-benzenetetracarboxylic acid diimide, 1,2,4,5-benzenetetracarboxylic 1,2:4,5-diimide, benzene-1,2,4,5-tetracarboxylic diimide, pyrrolo 3,4-f isoindole-1,3,5,7-tetrone, pyrrolo 3,4-f isoindole-1,3,5,7 2h,6h-tetrone
PubChem CID 75696
IUPAC Name pyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
SMILES C1=C2C(=CC3=C1C(=O)NC3=O)C(=O)NC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1437206
View Documents
Thermo Scientific Chemicals
L1437206
5 g
Each for $83.68
Only null left
 
AAL1437214
View Documents
Thermo Scientific Chemicals
L1437214
25 g
Each for $273.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2550-73-4
216.152
UGQZLDXDWSPAOM-UHFFFAOYSA-N
75696
C1=C2C(=CC3=C1C(=O)NC3=O)C(=O)NC2=O
C10H4N2O4
MFCD00005004
pyromellitic diimide, pyromellitimide, pyromellitic acid diimide, benzo 1,2-c:4,5-c' dipyrrole-1,3,5,7 2h,6h-tetrone, unii-32xa0w314g, 1,2,4,5-benzenetetracarboxylic acid diimide, 1,2,4,5-benzenetetracarboxylic 1,2:4,5-diimide, benzene-1,2,4,5-tetracarboxylic diimide, pyrrolo 3,4-f isoindole-1,3,5,7-tetrone, pyrrolo 3,4-f isoindole-1,3,5,7 2h,6h-tetrone
pyrrolo[3,4-f]isoindole-1,3,5,7-tetrone

Specifications

2550-73-4
C10H4N2O4
5 g
pyromellitic diimide, pyromellitimide, pyromellitic acid diimide, benzo 1,2-c:4,5-c' dipyrrole-1,3,5,7 2h,6h-tetrone, unii-32xa0w314g, 1,2,4,5-benzenetetracarboxylic acid diimide, 1,2,4,5-benzenetetracarboxylic 1,2:4,5-diimide, benzene-1,2,4,5-tetracarboxylic diimide, pyrrolo 3,4-f isoindole-1,3,5,7-tetrone, pyrrolo 3,4-f isoindole-1,3,5,7 2h,6h-tetrone
C1=C2C(=CC3=C1C(=O)NC3=O)C(=O)NC2=O
216.152
216.15
Pyromellitic diimide
>300°C
MFCD00005004
214780
UGQZLDXDWSPAOM-UHFFFAOYSA-N
pyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
75696
97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-852-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only