missing translation for 'onlineSavingsMsg'
Learn More

Pyromellitic diimide, 97%

Catalog No. AAL1437206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1437206 5 g
AAL1437214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1437206 Supplier Thermo Scientific Chemicals Supplier No. L1437206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2550-73-4 | C10H4N2O4 | 216.152 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2550-73-4
Molecular Formula C10H4N2O4
Molecular Weight (g/mol) 216.152
MDL Number MFCD00005004
InChI Key UGQZLDXDWSPAOM-UHFFFAOYSA-N
Synonym pyromellitic diimide, pyromellitimide, pyromellitic acid diimide, benzo 1,2-c:4,5-c' dipyrrole-1,3,5,7 2h,6h-tetrone, unii-32xa0w314g, 1,2,4,5-benzenetetracarboxylic acid diimide, 1,2,4,5-benzenetetracarboxylic 1,2:4,5-diimide, benzene-1,2,4,5-tetracarboxylic diimide, pyrrolo 3,4-f isoindole-1,3,5,7-tetrone, pyrrolo 3,4-f isoindole-1,3,5,7 2h,6h-tetrone
PubChem CID 75696
IUPAC Name pyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
SMILES C1=C2C(=CC3=C1C(=O)NC3=O)C(=O)NC2=O

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 214780
Formula Weight 216.15
Percent Purity 97%
Chemical Name or Material Pyromellitic diimide
EINECSNumber 219-852-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only