missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Pyrogallol | |
| 133°C | |
| 309°C | |
| MFCD00002192 | |
| 2811 | |
| WQGWDDDVZFFDIG-UHFFFAOYSA-N | |
| benzene-1,2,3-triol | |
| 1057 | |
| 126.11 | |
| Crystalline Powder |
| 87-66-1 | |
| White | |
| C6H6O3 | |
| 500 g | |
| pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral | |
| OC1=CC=CC(O)=C1O | |
| 126.11 | |
| CHEBI:16164 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 87-66-1 | |
| 126.11 | |
| WQGWDDDVZFFDIG-UHFFFAOYSA-N | |
| 1057 | |
| benzene-1,2,3-triol |
| C6H6O3 | |
| MFCD00002192 | |
| pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral | |
| CHEBI:16164 | |
| OC1=CC=CC(O)=C1O |
Safety and Handling
EINECSNumber : (3)-0554
RTECSNumber : UX2800000
TSCA : Yes