missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol 99.0+%, TCI America™

Catalog No. p-7134682
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P057025G 25 g
P0570100G 100 g
P0570500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P057025G Supplier TCI America Supplier No. P057025G
Only null left

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
PubChem CID 1057
ChEBI CHEBI:16164
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O

Specifications

Melting Point 133°C
Color White
Boiling Point 309°C
Quantity 25 g
UN Number 2811
Formula Weight 126.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pyrogallol
EINECSNumber (3)-0554
RTECSNumber UX2800000
TSCA Yes