missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrimidine free base 98.0+%, TCI America™
$57.79 - $160.30
Chemical Identifiers
| CAS | 289-95-2 |
|---|---|
| Molecular Formula | C4H4N2 |
| Molecular Weight (g/mol) | 80.09 |
| MDL Number | MFCD00006059 |
| InChI Key | CZPWVGJYEJSRLH-UHFFFAOYSA-N |
| Synonym | 1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l |
| PubChem CID | 9260 |
| ChEBI | CHEBI:16898 |
| IUPAC Name | pyrimidine |
| SMILES | C1=CN=CN=C1 |
Chemical Identifiers
| 289-95-2 | |
| 80.09 | |
| CZPWVGJYEJSRLH-UHFFFAOYSA-N | |
| 9260 | |
| pyrimidine |
| C4H4N2 | |
| MFCD00006059 | |
| 1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l | |
| CHEBI:16898 | |
| C1=CN=CN=C1 |
Specifications
| 289-95-2 | |
| 123°C | |
| MFCD00006059 | |
| 1325 | |
| CZPWVGJYEJSRLH-UHFFFAOYSA-N | |
| pyrimidine | |
| 9260 | |
| 80.09 | |
| Crystalline Lumps |
| White | |
| C4H4N2 | |
| 1 g | |
| 1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l | |
| C1=CN=CN=C1 | |
| 80.09 | |
| CHEBI:16898 | |
| ≥98.0% (GC,T) | |
| Pyrimidine free base |
Safety and Handling
RTECSNumber : UV6263000
TSCA : Yes