accessibility menu, dialog, popup

UserName

NN

CAS RN 289-95-2

IUPAC Name: pyrimidine
Synonyms: pyrimidine miazine dimer pyrimidine PYR metadiazine M-diazine pyrimidine base pyrimidine dimer pyrimidine, dimer
Molecular Weight (g/mol): 80.09
Molecular Formula: C4H4N2
InChi Key: CZPWVGJYEJSRLH-UHFFFAOYSA-N
SMILES: C1=CN=CN=C1
Boiling Point 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)