missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine, 99%

CAS: 289-95-2 | C4H4N2 | 80.09 g/mol

$207.92 - $207.92

Chemical Identifiers

CAS 289-95-2
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006059
InChI Key CZPWVGJYEJSRLH-UHFFFAOYSA-N
Synonym 1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l
PubChem CID 9260
ChEBI CHEBI:16898
IUPAC Name pyrimidine
SMILES C1=CN=CN=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC157700050
SDS View Documents
Thermo Scientific Chemicals
157700050
5 g Glass bottle
Each for $207.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

289-95-2
80.09
CZPWVGJYEJSRLH-UHFFFAOYSA-N
9260
pyrimidine
C4H4N2
MFCD00006059
1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l
CHEBI:16898
C1=CN=CN=C1

Specifications

289-95-2
Colorless to Orange
123°C to 124°C
Authentic
Glass bottle
1.5010 to 1.504
5 g
1.01
1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l
CZPWVGJYEJSRLH-UHFFFAOYSA-N
pyrimidine
9260
80.09
Liquid
20°C to 22°C
1.0100g/mL
31°C
98.5% min. (GC)
C4H4N2
MFCD00006059
23, 89
15, 8100
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
C1=CN=CN=C1
80.09
CHEBI:16898
99%
Pyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-026-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only