Learn More
Pyrimidine, 98+%
CAS: 289-95-2 | C4H4N2 | 80.09 g/mol
Supplier: Thermo Scientific Chemicals A1553406
Description
Used in the synthesis and discovery of antiviral medication, such as in the case of HIV and HSV. Also used in the synthesis of potent inhibitors of 15-lipoxygenase in the reduction of the release of leukotrienes. And in ithe production of RNA and DNA, proteins and starches, the regulation of enzymes and cell signaling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in the synthesis and discovery of antiviral medication, such as in the case of HIV and HSV. Also used in the synthesis of potent inhibitors of 15-lipoxygenase in the reduction of the release of leukotrienes. And in ithe production of RNA and DNA, proteins and starches, the regulation of enzymes and cell signaling.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Specifications
| Pyrimidine | |
| 20°C to 22°C | |
| 123°C to 124°C | |
| C4H4N2 | |
| MFCD00006059 | |
| UN1993 | |
| Hygroscopic | |
| 1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l | |
| CZPWVGJYEJSRLH-UHFFFAOYSA-N | |
| pyrimidine | |
| 9260 | |
| 80.09 |
| 289-95-2 | |
| 1.016 | |
| 31°C (87°F) | |
| 1.504 | |
| 5 g | |
| 103894 | |
| 14,7987 | |
| Fully miscible in water. | |
| C1=CN=CN=C1 | |
| 80.09 | |
| CHEBI:16898 | |
| 99% |
Chemical Identifiers
| 289-95-2 | |
| 80.09 | |
| CZPWVGJYEJSRLH-UHFFFAOYSA-N | |
| 9260 | |
| pyrimidine |
| C4H4N2 | |
| MFCD00006059 | |
| 1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l | |
| CHEBI:16898 | |
| C1=CN=CN=C1 |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 206-026-0
RTECSNumber : UV6263000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only