missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-carboxylic acid, 95%

CAS: 4595-61-3 | C5H4N2O2 | 124.10 g/mol

Supplier:  Thermo Scientific Chemicals H6140606

 View more versions of this product

Catalog No. AAH6140606


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pyrimidine-5-carboxylic acid
259°C to 261°C
MFCD00856162
5-pyrimidinecarboxylic acid, 5-carboxypyrimidine, pyrimidine-5-carboxilic acid, 5-pyrimidine carboxylic acid, pyrimidine-5-carboxylate acid, pyrimidine-5-carboxylicacid, pubchem2056, acmc-209k5a, 5-pyrimidyl carboxylic acid, pyrimidin-5-carboxylic acid
OC(=O)C1=CN=CN=C1
124.10
124.1
Solid
4595-61-3
C5H4N2O2
5 g
IIVUJUOJERNGQX-UHFFFAOYSA-N
pyrimidine-5-carboxylic acid
78346
95%

Chemical Identifiers

4595-61-3
124.10
IIVUJUOJERNGQX-UHFFFAOYSA-N
78346
OC(=O)C1=CN=CN=C1
C5H4N2O2
MFCD00856162
5-pyrimidinecarboxylic acid, 5-carboxypyrimidine, pyrimidine-5-carboxilic acid, 5-pyrimidine carboxylic acid, pyrimidine-5-carboxylate acid, pyrimidine-5-carboxylicacid, pubchem2056, acmc-209k5a, 5-pyrimidyl carboxylic acid, pyrimidin-5-carboxylic acid
pyrimidine-5-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-994-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only