missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-boronic acid hemihydrate, 97%

CAS: 696602-91-2 | C8H12B2N4O5 | 265.827 g/mol

$195.95 - $582.55

Chemical Identifiers

CAS 696602-91-2
Molecular Formula C8H12B2N4O5
Molecular Weight (g/mol) 265.827
MDL Number MFCD03002366
InChI Key YXMKGFBYXBTOJS-UHFFFAOYSA-N
Synonym pyrimidin-5-ylboronic acid hemihydrate, pyrimidine-5-boronic acid hemihydrate, 132.91 anhydrous, c4h5bn2o2.5h2o, bis pyrimidin-5-ylboronic acid hydrate
PubChem CID 67112167
IUPAC Name pyrimidin-5-ylboronic acid;hydrate
SMILES B(C1=CN=CN=C1)(O)O.B(C1=CN=CN=C1)(O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2921203
View Documents
Thermo Scientific Chemicals
H2921203
1 g N/A
Only null left
 
AAH2921209
View Documents
Thermo Scientific Chemicals
H2921209
10 g N/A
Only null left
 
Description

Description

It is employed as a intermediates for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

696602-91-2
265.827
YXMKGFBYXBTOJS-UHFFFAOYSA-N
67112167
B(C1=CN=CN=C1)(O)O.B(C1=CN=CN=C1)(O)O.O
C8H12B2N4O5
MFCD03002366
pyrimidin-5-ylboronic acid hemihydrate, pyrimidine-5-boronic acid hemihydrate, 132.91 anhydrous, c4h5bn2o2.5h2o, bis pyrimidin-5-ylboronic acid hydrate
pyrimidin-5-ylboronic acid;hydrate

Specifications

696602-91-2
C8H12B2N4O5
1 g
YXMKGFBYXBTOJS-UHFFFAOYSA-N
pyrimidin-5-ylboronic acid;hydrate
67112167
97%
>300°C
MFCD03002366
pyrimidin-5-ylboronic acid hemihydrate, pyrimidine-5-boronic acid hemihydrate, 132.91 anhydrous, c4h5bn2o2.5h2o, bis pyrimidin-5-ylboronic acid hydrate
B(C1=CN=CN=C1)(O)O.B(C1=CN=CN=C1)(O)O.O
265.827
132.91 (123.91 Anhydrous)
Pyrimidine-5-boronic acid hemihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only