CAS RN 696602-91-2
IUPAC Name:
bis((pyrimidin-5-yl)boronic acid) hydrate
Synonyms:
bis(pyrimidin-5-ylboronic acid) hydrate
Molecular Weight (g/mol):
265.83
Molecular Formula:
C8H12B2N4O5
InChi Key:
YXMKGFBYXBTOJS-UHFFFAOYSA-N
SMILES:
O.OB(O)C1=CN=CN=C1.OB(O)C1=CN=CN=C1
1
–
1
of
1
results
1
Promotions Available
Pyrimidine-5-boronic acid hemihydrate, 97%
Pyrimidine-5-boronic acid hemihydrate, 97%
CAS: 696602-91-2 Molecular Formula: C8H12B2N4O5 Molecular Weight (g/mol): 265.827 MDL Number: MFCD03002366 InChI Key: YXMKGFBYXBTOJS-UHFFFAOYSA-N Synonym: pyrimidin-5-ylboronic acid hemihydrate,pyrimidine-5-boronic acid hemihydrate,132.91 anhydrous,c4h5bn2o2.5h2o,bis pyrimidin-5-ylboronic acid hydrate PubChem CID: 67112167 IUPAC Name: pyrimidin-5-ylboronic acid;hydrate SMILES: B(C1=CN=CN=C1)(O)O.B(C1=CN=CN=C1)(O)O.O