missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-boronic acid hemihydrate, 97%

Catalog No. AAH2921203
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2921203 1 g
AAH2921209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2921203 Supplier Thermo Scientific Chemicals Supplier No. H2921203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 696602-91-2 | C8H12B2N4O5 | 265.827 g/mol

It is employed as a intermediates for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 696602-91-2
Molecular Formula C8H12B2N4O5
Molecular Weight (g/mol) 265.827
MDL Number MFCD03002366
InChI Key YXMKGFBYXBTOJS-UHFFFAOYSA-N
Synonym pyrimidin-5-ylboronic acid hemihydrate, pyrimidine-5-boronic acid hemihydrate, 132.91 anhydrous, c4h5bn2o2.5h2o, bis pyrimidin-5-ylboronic acid hydrate
PubChem CID 67112167
IUPAC Name pyrimidin-5-ylboronic acid;hydrate
SMILES B(C1=CN=CN=C1)(O)O.B(C1=CN=CN=C1)(O)O.O

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 132.91 (123.91 Anhydrous)
Percent Purity 97%
Chemical Name or Material Pyrimidine-5-boronic acid hemihydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only