missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-2-thiocarboxamide, 97%

Catalog No. p-7036481
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5478303 1 g
AAH5478306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5478303 Supplier Thermo Scientific Chemicals Supplier No. H5478303
Only null left

CAS: 4537-73-9 | C5H5N3S | 139.176 g/mol

Pyrimidine-2-thiocarboxamide is used as a pharmaceutical intermediate and as a synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrimidine-2-thiocarboxamide is used as a pharmaceutical intermediate and as a synthetic building block.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 4537-73-9
Molecular Formula C5H5N3S
Molecular Weight (g/mol) 139.176
MDL Number MFCD12093703
InChI Key QAUHVPUYFSGVME-UHFFFAOYSA-N
Synonym 2-pyrimidinecarbothioamide, pyrimidine-2-thiocarboxamide, 2-pyrimidinecarbothioamide 9ci
PubChem CID 23273397
IUPAC Name pyrimidine-2-carbothioamide
SMILES C1=CN=C(N=C1)C(=S)N

Specifications

Quantity 1 g
Beilstein 508162
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material Pyrimidine-2-thiocarboxamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only