missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-2-sulfonyl Fluoride 98.0+%, TCI America™

Catalog No. P24651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24651G 1 g
P24655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24651G Supplier TCI America Supplier No. P24651G
Only null left

Chemical Identifiers

CAS 35762-87-9
Molecular Formula C4H3FN2O2S
Molecular Weight (g/mol) 162.14
MDL Number MFCD09265503
InChI Key HMZIGHBQOOGZAK-UHFFFAOYSA-N
PubChem CID 12175574
IUPAC Name pyrimidine-2-sulfonyl fluoride
SMILES FS(=O)(=O)C1=NC=CC=N1

Specifications

Melting Point 60°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 162.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pyrimidine-2-sulfonyl Fluoride
TSCA No
Recommended Storage Refrigerator