missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. P23281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23281G Supplier TCI America Supplier No. P23281G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31519-62-7
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.10
MDL Number MFCD00856161
InChI Key ZFCHNZDUMIOWFV-UHFFFAOYSA-N
Synonym 2-pyrimidinecarboxylic acid, 2-carboxypyrimidine, pyrimidine carboxylic acid, pyrimidinecarboxylic acid, 2-carboxy-1,3-diazine, 2-pyrimidinecarboxylic acid 6ci,8ci,9ci, 2-pyrimidinecarboxylicacid, 2-carboxy pyrimidine, pubchem9603, pyrimidine-carboxylic acid
PubChem CID 12626245
IUPAC Name pyrimidine-2-carboxylic acid
SMILES OC(=O)C1=NC=CC=N1

Specifications

Melting Point 199°C
Color White
Quantity 1 g
Formula Weight 124.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrimidine-2-carboxylic Acid