accessibility menu, dialog, popup

UserName

CAS RN 31519-62-7

IUPAC Name: pyrimidine-2-carboxylic acid
Synonyms: pyrimidine-2-carboxylic acid
Molecular Weight (g/mol): 124.1
Molecular Formula: C5H4N2O2
InChi Key: ZFCHNZDUMIOWFV-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=CC=N1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)