Learn More
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific™
$113.64 - $342.89
Chemical Identifiers
| CAS | 31519-62-7 |
|---|---|
| Molecular Formula | C5H4N2O2 |
| Molecular Weight (g/mol) | 124.10 |
| MDL Number | MFCD00856161 |
| InChI Key | ZFCHNZDUMIOWFV-UHFFFAOYSA-N |
| Synonym | 2-pyrimidinecarboxylic acid, 2-carboxypyrimidine, pyrimidine carboxylic acid, pyrimidinecarboxylic acid, 2-carboxy-1,3-diazine, 2-pyrimidinecarboxylic acid 6ci,8ci,9ci, 2-pyrimidinecarboxylicacid, 2-carboxy pyrimidine, pubchem9603, pyrimidine-carboxylic acid |
| PubChem CID | 12626245 |
| IUPAC Name | pyrimidine-2-carboxylic acid |
| SMILES | OC(=O)C1=NC=CC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC455830010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC455830050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 31519-62-7 | |
| 124.10 | |
| ZFCHNZDUMIOWFV-UHFFFAOYSA-N | |
| 12626245 | |
| OC(=O)C1=NC=CC=N1 |
| C5H4N2O2 | |
| MFCD00856161 | |
| 2-pyrimidinecarboxylic acid, 2-carboxypyrimidine, pyrimidine carboxylic acid, pyrimidinecarboxylic acid, 2-carboxy-1,3-diazine, 2-pyrimidinecarboxylic acid 6ci,8ci,9ci, 2-pyrimidinecarboxylicacid, 2-carboxy pyrimidine, pubchem9603, pyrimidine-carboxylic acid | |
| pyrimidine-2-carboxylic acid |
Specifications
| 31519-62-7 | |
| 94% min. (HPLC) | |
| C5H4N2O2 | |
| 2-pyrimidinecarboxylic acid, 2-carboxypyrimidine, pyrimidine carboxylic acid, pyrimidinecarboxylic acid, 2-carboxy-1,3-diazine, 2-pyrimidinecarboxylic acid 6ci,8ci,9ci, 2-pyrimidinecarboxylicacid, 2-carboxy pyrimidine, pubchem9603, pyrimidine-carboxylic acid | |
| OC(=O)C1=NC=CC=N1 | |
| 124.10 | |
| 124.1 | |
| Pyrimidine-2-carboxylic acid |
| Authentic | |
| Glass Bottle | |
| MFCD00856161 | |
| ZFCHNZDUMIOWFV-UHFFFAOYSA-N | |
| pyrimidine-2-carboxylic acid | |
| 12626245 | |
| 95% |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
RUO – Research Use Only