missing translation for 'onlineSavingsMsg'
Learn More

Pyrimethamine 98.0+%, TCI America™

Catalog No. P20371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20371G 1 g
P20375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20371G Supplier TCI America Supplier No. P20371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58-14-0
Molecular Formula C12H13ClN4
Molecular Weight (g/mol) 248.71
MDL Number MFCD00057350
InChI Key WKSAUQYGYAYLPV-UHFFFAOYSA-N
Synonym pyrimethamine, chloridine, daraprim, chloridin, diaminopyritamin, 5-4-chlorophenyl-6-ethylpyrimidine-2,4-diamine, ethylpyrimidine, pirimetamina, chloridyn, malocide
PubChem CID 4993
ChEBI CHEBI:8673
IUPAC Name 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
SMILES CCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1

Specifications

Melting Point 241°C
Color White
Quantity 1 g
UN Number 3077
Formula Weight 248.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrimethamine
EINECSNumber (9)-0539&(9)-0696&(9)-1124&(9)-0270
RTECSNumber UV8140000
TSCA No