missing translation for 'onlineSavingsMsg'
Learn More

Pyrimethamine, 98%, Thermo Scientific Chemicals

$65.93 - $65.93

Chemical Identifiers

CAS 58-14-0
Molecular Formula C12H13ClN4
Molecular Weight (g/mol) 248.71
MDL Number MFCD00057350
InChI Key WKSAUQYGYAYLPV-UHFFFAOYSA-N
Synonym pyrimethamine, chloridine, daraprim, chloridin, diaminopyritamin, 5-4-chlorophenyl-6-ethylpyrimidine-2,4-diamine, ethylpyrimidine, pirimetamina, chloridyn, malocide
PubChem CID 4993
ChEBI CHEBI:8673
IUPAC Name 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
SMILES CCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC461200010
View Documents
Thermo Scientific Chemicals
461200010
1 g
Each for $65.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

58-14-0
248.71
WKSAUQYGYAYLPV-UHFFFAOYSA-N
4993
5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
C12H13ClN4
MFCD00057350
pyrimethamine, chloridine, daraprim, chloridin, diaminopyritamin, 5-4-chlorophenyl-6-ethylpyrimidine-2,4-diamine, ethylpyrimidine, pirimetamina, chloridyn, malocide
CHEBI:8673
CCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1

Specifications

58-14-0
0.5% max. (105°C, 4 h)
97.5% min. (HPLC)
C12H13ClN4
1 g
Solubility in water: insoluble.
CCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1
248.71
CHEBI:8673
98%
241.0°C
Authentic
Glass Bottle
MFCD00057350
pyrimethamine, chloridine, daraprim, chloridin, diaminopyritamin, 5-4-chlorophenyl-6-ethylpyrimidine-2,4-diamine, ethylpyrimidine, pirimetamina, chloridyn, malocide
WKSAUQYGYAYLPV-UHFFFAOYSA-N
5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
4993
248.71
Pyrimethamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed. Suspected of damaging fertility or the unborn child.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 200-364-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only