missing translation for 'onlineSavingsMsg'
Learn More

Pyrimethamine, 98%, Thermo Scientific Chemicals

Catalog No. AC461200010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461200010 1 g
AC461200050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461200010 Supplier Thermo Scientific Chemicals Supplier No. 461200010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-14-0
Molecular Formula C12H13ClN4
Molecular Weight (g/mol) 248.71
MDL Number MFCD00057350
InChI Key WKSAUQYGYAYLPV-UHFFFAOYSA-N
Synonym pyrimethamine, chloridine, daraprim, chloridin, diaminopyritamin, 5-4-chlorophenyl-6-ethylpyrimidine-2,4-diamine, ethylpyrimidine, pirimetamina, chloridyn, malocide
PubChem CID 4993
ChEBI CHEBI:8673
IUPAC Name 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
SMILES CCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1

Specifications

Melting Point 241.0°C
Loss on Drying 0.5% max. (105°C, 4 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 248.71
Percent Purity 98%
Chemical Name or Material Pyrimethamine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 200-364-2

RUO – Research Use Only