missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxine Impurity A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 5196-20-3 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C8H9 NO2 |
| Synonym | 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol; 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 151.16 |
| Grade | Certified Reference Material |