missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxamine dihydrochloride, 98%

Catalog No. p-4228896
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436250010 1 g Glass bottle
AC436250050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436250010 Supplier Thermo Scientific Chemicals Supplier No. 436250010

CAS: 524-36-7 | C8H12N2O2·2ClH | 241.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 524-36-7
Molecular Formula C8H12N2O2·2ClH
Molecular Weight (g/mol) 241.12
InChI Key HNWCOANXZNKMLR-UHFFFAOYSA-N
Synonym pyridoxamine dihydrochloride, 4-aminomethyl-5-hydroxymethyl-2-methylpyridin-3-ol dihydrochloride, pyridoxamine dichlorohydrate, pyridoxylamine dihydrochloride, 4-aminomethyl-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride, unii-yq8ncr7v0o, pyridoxamine 2hcl, yq8ncr7v0o, 2-methyl-3-hydroxy-4-aminomethyl-5-hydroxymethylpyridene dihydrochloride, pyridoxemine, dihydrochloride
PubChem CID 10664
IUPAC Name 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;dihydrochloride
SMILES CC1=NC=C(C(=C1O)CN)CO.Cl.Cl

Specifications

Melting Point 216.0°C to 218.0°C
Color Brown to White
Elemental Analysis 36.15 to 38.01% C, 10.54 to 11.08%N (based on monohydrate)
Infrared Spectrum Authentic
Assay Percent Range 97.5 to max. 102.5% min. (on dry basis) (Argentometry)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,8093
Formula Weight 241.12
Percent Purity 98%
Physical Form Powder or Solid
Chemical Name or Material Pyridoxamine dihydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 208-357-6

RUO – Research Use Only