missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxal Hydrochloride 98.0+%, TCI America™
$46.60 - $303.84
Chemical Identifiers
| CAS | 65-22-5 |
|---|---|
| Molecular Formula | C8H10ClNO3 |
| Molecular Weight (g/mol) | 203.622 |
| MDL Number | MFCD00012809 |
| InChI Key | FCHXJFJNDJXENQ-UHFFFAOYSA-N |
| Synonym | pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride |
| PubChem CID | 6171 |
| IUPAC Name | 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride |
| SMILES | CC1=NC=C(C(=C1O)C=O)CO.Cl |
Chemical Identifiers
| 65-22-5 | |
| 203.622 | |
| FCHXJFJNDJXENQ-UHFFFAOYSA-N | |
| 6171 | |
| CC1=NC=C(C(=C1O)C=O)CO.Cl |
| C8H10ClNO3 | |
| MFCD00012809 | |
| pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride | |
| 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride |
Specifications
| 65-22-5 | |
| C8H10ClNO3 | |
| 1 g | |
| FCHXJFJNDJXENQ-UHFFFAOYSA-N | |
| 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride | |
| 6171 | |
| ≥98.0% (HPLC,T) | |
| Pyridoxal Hydrochloride |
| White-Yellow | |
| MFCD00012809 | |
| pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride | |
| CC1=NC=C(C(=C1O)C=O)CO.Cl | |
| 203.622 | |
| 203.62 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1040
RTECSNumber : UV1225000
TSCA : Yes