accessibility menu, dialog, popup

UserName

HClCH3NOHOHO

CAS RN 65-22-5

IUPAC Name: hydrogen 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde chloride
Synonyms: hydrogen pyridoxal chloride
Molecular Weight (g/mol): 203.62
Molecular Formula: C8H10ClNO3
InChi Key: FCHXJFJNDJXENQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=NC=C(CO)C(C=O)=C1O
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (3)
  • (8)
Quantity 
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (5)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (8)
  • (5)
  • (6)
  • (3)
  • (8)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (5)
  • (3)